Light Leaper Accessory
# de catalogue 470320-722
Fournisseur: United Scientific Supplies
Spécifications
- Description:Light leaper accessory
- Cat. no.:470320-722
Spécifications
A propos de cet article
Vividly illustrate electrical charges.
- Easy to set up design
- Works with Wimshurst or Van de Graff generators
This is a popular accessory for electrostatic demonstrations. An insulating printed circuit board carries a metallic conduction path with six breaks and a binding post at each end.
When connected to an operating electrostatic generator, simultaneous sparks occur in each of the breaks as charge flows between the poles of the generator.
Customers Who Bought This Also Bought