DURAN® Aspirator (Levelling) Bottles, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Complete with screw connection cap, silicone seal and stopcock with PTFE spindle. Dosing of liquids is possible via a stopcock.
- Neck with DIN thread GL 45
- Tabulator with GL 32
- Retrace code
- Autoclavable
Customers Who Bought This Also Bought