WHEATON® Replacement Impellers for Magna Flex Spinner Flask, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Replacement impellers for magna flex spinner flask.
- Designed for microcarrier cultures with a magnetic stirrer
- Gentle stirring action provided by a bulb-shaped glass impeller
Customers Who Bought This Also Bought