Cryo-LazrTAG™, Hagaki Size Permanent Cryogenic Labels for Laser Printers, GA International
Fournisseur: GA INTERNATIONAL INC
Certificats
A propos de cet article
These labels can withstand deep-freezing and PCR thermocycler temperatures.
- Temperature range: –196 to 120 °C (–321 to 248 °F)
- Multiple freeze-thaw cycles
Cryo-LazrTAG™ cryogenic plastic rectangle labels are designed for use with 0.2 ml PCR tubes, 0.5, 1.5, and 2 ml microcentrifuge tubes, and 1 to 2 ml cryo vials.
Customers Who Bought This Also Bought