Stopper, Spherical Penny Head, Chemscience
# de catalogue CA10202-936
Fournisseur: CHEM SCIENCE INC CA
Spécifications
- Description:Stopper, spherical, penny head
- Cat. no.:CA10202-936
- Joint Size:35
- Supplier No.:SI-S0450035
Spécifications
A propos de cet article
Hollow stopper for use with spherical socket joint.
Customers Who Bought This Also Bought