VWR® Shelves for Reagent Rack Island Shelving System
Supplier: VWR International
Certificates
About this item
VWR® Reagent Rack systems provide excellent storage of laboratory equipment above the lab station.
- Max. load capacity of 176 lbs.
- Light neutral or snow white
Frequently Bought Together