A propos de cet article
Superior heat resistance with complete protection for the hand and wrist.
- Nonwoven felt insulation and a full-length heat barrier designed for intermittent handling of dry objects up to 200 °C (400 °F)
- Two-piece construction eliminates out seams for better finger comfort, dexterity, and flexibility
- Bonded nitrile coating protects against snags, punctures, abrasions, and cuts
- Provides excellent grip when handling dry or oily surfaces
- Contains no asbestos
- Compliant to FDA food handling requirements
For the best protection and comfort in moderate heat applications, ActivArmr® 42-474 gloves are the answer. Their nonwoven felt insulation and full-length heat barrier protect the worker, while two-piece construction eliminates out seams to provide better finger comfort, dexterity, and flexibility. These gloves feature an exclusive bonded nitrile coating to help protect against snags, punctures, abrasions, and cuts, while also enhancing dry grip. ActivArmr® 42-474 gloves wear an average of four times longer than 24 ounce cotton hot mill gloves. These gloves were designed to handle intermittent handling of hot cores, castings, and controls up to 400 °F, not as a replacement for silicate gloves.
Spécifications
- Catalog No:
- 76376-598
- 76376-600
- 76376-602
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Liner material:
- Felt
- Felt
- Felt
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet