Order Entry
ContactUsLinkComponent
Difco™ OGYE Agar Base, BD
Difco™ OGYE Agar Base, BD
# de catalogue CA90000-980
Fournisseur:  BD
Numéro CAS:  
Difco™ OGYE Agar Base, BD
# de catalogue CA90000-980
Fournisseur:  BD
Numéro CAS:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Spécifications

  • Description:
    Oxytetracycline glucose yeast extract agar base
  • Size:
    500 g
  • Cat. no.:
    CA90000-980
  • Supplier No.:
    218111

Spécifications

A propos de cet article

Oxytetracycline glucose yeast extract is used with antimicrobic vial oxytetracycline for selective isolation of yeasts and molds in foods.

  • Manufactured in an animal origin-free, antibiotic-free facility
  • Mycoplasma tested
  • Endotoxin tested

The individual ingredients and final dehydrated product are carefully quality controlled to assure consistent, high-quality performance and obviate the need for media to be prepared in the laboratory from raw materials.

Customers Who Bought This Also Bought