Manifolds, Nitrogen Argon Line, PTFE, Chemscience, SATI International Scientific
Fournisseur: CHEM SCIENCE INC CA
Certificats
A propos de cet article
These manifolds have a 14/20 outer joint at the center to fit a rubber septum for keeping syringes under an inert environment cutting down on the number used.
- Main tube has 22 mm O.D. with 10 mm hose connection
Delivery ports have 8 mm hose connections.
Customers Who Bought This Also Bought