KIMBLE® Atmospheric Environmental Bottles, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
The design of the atmospheric environmental bottle is recommended by the Atmospheric Environment Service (AES).
- KimCote® plastic safety coating
- Borosilicate glass high vacuum plug valve stems instead of PTFE stems, eliminating the chance of outgassing
- FKM valve stem o-rings that are easily changed
- Inlet stems ground to ¹/₂" Ext.Ø
- Manufactured from 33 expansion, low extractable borosilicate glass
Available in either a single or double valve configuration.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought