KIMBLE® Distilling Flasks, Round Bottom, 3-Neck, DWK Life Sciences
Fournisseur: Avantor
Certificats
A propos de cet article
Indented three-neck round bottom flasks with standard taper outer joints, all vertical.
- Heavy wall
- Indentations enhance agitation when stirring
- Manufactured from 33 expansion, low extractable borosilicate glass
This flask is also referred to as a Morton flask.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought