KIMBLE® Distilling Flasks, Round Bottom, 5-Neck, DWK Life Sciences
# de catalogue KT607500-1024
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Capacity:2000 ml
- Description:Distilling flask
- Cat. no.:KT607500-1024
- ST Joint Size:24/40 center; 24/40 side
- Supplier No.:607500-1024
Spécifications
A propos de cet article
Five-neck heavy wall round-bottom flask with vertical standard taper outer joints.
- Side necks are 90° apart
- Where possible, vertical necks will be supplied
- Manufactured from 33 expansion, low extractable borosilicate glass
On small flasks having large necks, positioning may have to be made at an angle.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought