KIMBLE® Foam-Type Distillation Trap, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Placed above a boiling flask to prevent foaming-type reactions from entering either the distilling column or the condenser.
- Features a more extensive baffle system than other traps
- Manufactured from 33 expansion, low extractable borosilicate glass
Useful with rotary evaporators.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought