KIMBLE® Petrochemical Filter Stick Apparatus with Hooks and Springs, DWK Life Sciences
# de catalogue KT513880-0000
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Filter stick apparatus
- Cat. no.:KT513880-0000
- ST Joint Size:24/40
- Supplier No.:513880-0000
Spécifications
A propos de cet article
Apparatus used in the determination of oil content and solvent extractables in petroleum waxes.
- Ref: ASTM D721
- Manufactured from 33 expansion, low extractable borosilicate glass
Assembly consists of a sintered glass filter stick with air pressure inlet tube and delivery nozzle and a cooling tube with 24/40 joints.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Delivery: Supplied with one pair of springs.
Customers Who Bought This Also Bought