KIMBLE® Distilling Flasks, Round Bottom, 3-Neck, DWK Life Sciences
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
A propos de cet article
Angled three neck round bottom flasks with standard taper outer joints.
- Side necks are angled
- Manufactured from 33 expansion, low extractable borosilicate glass
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought