KIMBLE® Distillation Receiver, Graduated Tube, DWK Life Sciences
Catalog # KT288251-0000
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Capacity:12 ml
- Description:Distillation receiver
- Length:142 mm
- Cat. no.:KT288251-0000
- ST Joint Size:14/20
- Graduations:0 - 2 ml
- Supplier No.:288251-0000
Specifications
About this item
This receiver is a component of the distillation apparatus.
Customers Who Bought This Also Bought