KIMBLE® Cow Type Distillation Receiver with Three Receivers and 14/20 [ST] Joints, DWK Life Sciences
# de catalogue KT287800-0000
Fournisseur: DWK Life Sciences (KIMBLE)
Spécifications
- Description:Distillation receiver
- Cat. no.:KT287800-0000
- ST Joint Size:14/20
- Supplier No.:287800-0000
Spécifications
A propos de cet article
Distillation receivers 14/20 cow type with three receivers.
- Cow-type design having three receivers spaced 45° apart
- Manufactured from 33 expansion, low extractable borosilicate glass
Condensers with drip joints will protrude into the spherical section, keeping wetted surfaces and product hold-up to a minimum.
Certifications: Borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Customers Who Bought This Also Bought