Miller® TurboLite™ T-BAK EXTREME Sharp Edge Personal Fall Limiters, Honeywell Safety
Fournisseur: Honeywell Safety Products
Certificats
A propos de cet article
Cable lifeline with web tie-back for sharp edge tie-back applications.
- Designed for use in sharp edge, tie-back applications with radius ≥0.005" (0.13 mm)
- Galvanized cable lifeline with Vectran™ web tie-back
- Available in lengths of 8 and 11'
- Single and twin configurations
Miller® TurboLite™ Personal Fall Limiters (PFLs) are designed for performance in edge applications, offering versatility, enhanced user mobility, and maximum safety. These devices maximize versatility for virtually any short-range application and provide protection for a wider range of workers. The quick and easy installation creates a better work experience while increasing safety and lowering the cost of ownership. These devices ensure compatibility with all Miller® harnesses. Equipped with an easy to install shock-absorbing connector system and galvanized cable lifeline with a web tie-back extension.
Certifications: Meets OSHA 1926.502, ANSI A10.32, ANSI Z359.14, Class B and LE (Leading Edge) requirements.
Customers Who Bought This Also Bought