Miller® TurboLite™ T-BAK MAX Smooth Edge Personal Fall Limiters, Honeywell Safety
Fournisseur: Honeywell Safety Products
Certificats
A propos de cet article
Web lifeline for smooth edge tie-back applications.
- Designed for use in smooth edge applications with radius ≥0.060" (1.5 mm)
- Abrasion resistant Vectran™ webbing tie-back lifeline
- Available in lengths of 7.5 and 10.5'
- Single and twin configurations
Miller® TurboLite™ personal fall limiters (PFLs) are designed for performance in edge applications, offering versatility, enhanced user mobility, and maximum safety. These devices maximize versatility for virtually any short-range application and provide protection for a wider range of workers. The quick and easy installation creates a better work experience while increasing safety and lowering the cost of ownership. These devices ensure compatibility with all Miller® harnesses. Equipped with an easy to install shock-absorbing connector system and abrasion-resistant Vectran™ webbing lifeline.
Certifications: Meets OSHA 1926.502, ANSI A10.32, and ANSI Z359.14, Class B requirements.
Customers Who Bought This Also Bought