Miller® TurboLite™ MAX Smooth Edge Personal Fall Limiters, Honeywell Safety
Fournisseur: Honeywell Safety Products
Certificats
A propos de cet article
Web lifeline for smooth edge applications.
- Designed for use in smooth edge applications with radius ≥ 0.060" (1.5 mm)
- Honeywell Spectra® webbing lifeline provides outstanding chemical, abrasion, and cut resistance
- Available in lengths of 6, 9 and 12'
- Single and twin configurations
- Variety of lifeline connector options
Miller® TurboLite™ Personal Fall Limiters (PFLs) are designed for performance in edge applications, offering versatility, enhanced user mobility, and maximum safety. These devices maximize versatility for virtually any short-range application and provide protection for a wider range of workers. The quick and easy installation creates a better work experience while increasing safety and lowering the cost of ownership. These devices ensure compatibility with all Miller® harnesses. It is equipped with an easy to install shock-absorbing connector system and abrasion-resistant Spectra® webbing lifeline.
Certifications: Meets OSHA 1926.502, ANSI A10.32, ANSI Z359.14, Class B and LE (Leading Edge), and CSA Z259.2.2 requirements.
Customers Who Bought This Also Bought