Miller® TurboLite™ EXTREME Sharp Edge Personal Fall Limiters, Honeywell Safety
Fournisseur: Honeywell Safety Products
Certificats
A propos de cet article
Cable lifeline for sharp edge applications.
- Designed for use in sharp edge applications with radius >0.005" (0.13 mm)
- Galvanized cable lifeline
- Available in lengths of 6 and 9'
- Single and twin configurations
- Variety of lifeline connector options
- Versatile, reliable, durable, simple, and safe
Miller® TurboLite™ personal fall limiters (PFLs) are designed for performance in edge applications, offering versatility, enhanced user mobility, and maximum safety. These devices maximize versatility for virtually any short-range application and provide protection for a wider range of workers. The quick and easy installation creates a better work experience while increasing safety and lowering the cost of ownership. These devices ensure compatibility with all Miller® harnesses. It is equipped with an easy to install shock-absorbing connector system and galvanized cable lifeline.
Customers Who Bought This Also Bought