VWR® Four Student Perimeter Workstation Made from Solid Oak
# de catalogue 89374-910
Fournisseur: VWR International
Spécifications
- No. of Users:4
- Worksurface Material:Epoxy resin
- Description:Book compartment table with cabinet
- Environmentally Preferable:Safer Human and Environmental Health
- Height:30"
- Environmental certification:MAS Certified Green
- Cat. no.:89374-910
- W×D:66×42"
Spécifications
A propos de cet article
Four student cabinet table constructed of solid oak, oak and hardwood veneers features a center cabinet with adjustable shelves, book compartments in the aprons, and locking doors.
- 1" solid epoxy resin surface
- Four student workstation features a locking center cabinet with adjustable shelf
- Includes four book compartments in the aprons
- The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
This table is available with plain or book compartment aprons and comes in both single (two-student) and double faced (four-student) models. The center cabinet is available with two locking drawers or as a plain base cabinet with adjustable shelf. These tables are designed for use at sitting height.
Certifications: It is SEFA compliant and MAS certified green.
Customers Who Bought This Also Bought