VWR® Four Student Cabinet Tables with Locking Drawers
Fournisseur: VWR International
Certificats
A propos de cet article
Four student cabinet table constructed of solid oak, oak and hardwood veneers features a center cabinet with adjustable shelves, locking doors and four locking drawers.
- 1¾" ChemGuard table surface
- Four student workstation features a locking center cabinet with adjustable shelf
- Includes four book compartments in the aprons
- The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
This table is available with plain or book compartment aprons and comes in both single (two-student) and double faced (four-student) models. The center cabinet is available with two locking drawers or as a plain base cabinet with adjustable shelf. These tables are designed for use at sitting height. This unit is available with our complete line of top materials to satisfy every requirement or budget.
Certifications: It is SEFA compliant and MAS certified green.
Spécifications
- Catalog No:
- 89374-892
- 89374-894
- 89374-884
- 89374-886
- 89374-908
- 89374-902
- 89374-900
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green