VWR® Four Student Cabinet Tables Construced of Solid Oak with Locking Doors and Drawers
Fournisseur: VWR International
Certificats
A propos de cet article
Four student cabinet table constructed of solid oak, oak and hardwood veneers features a center cabinet with adjustable shelves, locking doors and four locking drawers.
- ¾" phenolic resin surface
- Finishing the desk is rubber base molding, leg boots and a durable chemical resistant UV finish
- This cabinet table allows workspace and storage for four users
This table is available with plain or book compartment aprons and comes in both single (two-student) and double faced (four-student) models. These tables are designed for use at sitting height. This unit is available with our complete line of top materials to satisfy every requirement or budget.
Certifications: It is SEFA compliant and MAS certified green.
Spécifications
- Catalog No:
- 89374-896
- 89374-914
- 89374-888
- 89374-912
- 89374-890
- 89374-904
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green