VWR® Book Compartment Tables with Cabinet for Two Student
Fournisseur: VWR International
Certificats
A propos de cet article
Two student cabinet table constructed of solid oak, oak and hardwood veneers.
- 1" solid epoxy resin surface
- Includes two book compartments in the aprons
- The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
- Two student workstation features a locking center cabinet with two locking drawers
This table is available with plain or book compartment aprons and comes in both single (two-student) and double faced (four-student) models. The center cabinet is available with two locking drawers or as a plain base cabinet with adjustable shelf. These tables are designed for use at sitting height.
Certifications: It is SEFA compliant and MAS certified green.
Spécifications
- Catalog No:
- 89374-874
- 89374-880
- 89374-882
- 89374-858
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green