Brady® Safety Inspection Timers, Brady Worldwide
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
Shed some light on inspections by supplementing inspection tags with the inspection timer, and get an inspection status at a glance.
- Review and reset; after completing an inspection, reset the timer so it is ready for the next inspection
- Perfect for facilities that have internal audit and inspection schedules
- Sturdy ABS plastic construction
Unlike tags, the bright color LED lights provide an inspection status from a distance and break through the grime of any facility
IP 65 rated timers will survive harsh industrial environments, including exposure to water, dust, extreme temperatures and more.
Safety inspection timers are ideal for use in facilities that have multiple eyewash stations, emergency showers, emergency exit doors, exit signs, fire extinguishers, or first aid kits.
Certifications: Ingress Protection Rating IP 65.
Delivery: All timers are supplied with three AAA Alkaline batteries.
Customers Who Bought This Also Bought