Brady® BatteryBlock™ Power Connector Lockout - Forklift, Brady Worldwide
# de catalogue 76359-914
Fournisseur: Brady Worldwide
Spécifications
- Description:BatteryBlock™ power connector lockout
- Material:ABS plastic
- Cat. no.:76359-914
- Dimensions:5.25H×3.15W×2.5"D
Spécifications
A propos de cet article
For forklift safety, proper operation is a main concern, but a secure lockout can be just as important. Keep forklifts and lift vehicles locked out and locked down with the BatteryBlock™ power connector lockout.
- Device fits on 175 A and 350 A power connectors with and without handles
- Single-piece design is easy to store and manage so users don't have to worry about losing pieces
- Manage use and safety by applying two padlocks from authorized employees
Non-conductive pins ensures the device stays secure during maintenance projects and off-shift periods.
Certifications: OSHA 29 CFR 1910.147.
Customers Who Bought This Also Bought