Barcode Reader for Clarity Urine Analyzer, Clarity Diagnostics
# de catalogue 95064-148
Fournisseur: Clarity Diagnostics
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Description:Bar Code Reader
- Cat. no.:95064-148
- Supplier No.:DTG-UABC
Spécifications
A propos de cet article
Clarity barcode reader for use with Clarity Urocheck and Clarity Urocheck 120C analyzer.
- Connect through RS232 port
- Supplied with splitter cable
- Limited 1 year warranty
Ideal for use in hospitals.
Customers Who Bought This Also Bought