Bizwell Iona FR Coveralls, Portwest
Fournisseur: Portwest
Certificats
A propos de cet article
Coveralls are designed to protect against radiant, convective, and contact heat.
- Pockets: two front, two back, one concealed cell phone pocket, two side, two knee, and one rule
- Highly durable, flame resistant reflective tape
- Dual hazard protection
- ARC2
Certified protection against molten metal splash and guaranteed flame resistance for the life of the garment.
Certifications: EN ISO 11612 (A1+A2. B1. C1. E1. F1). EN ISO 11611 Class 1 (A1+A2). NFPA® 70E. ASTM F1506-10a. NFPA® 2112. ASTM F1959/F1959M-12 (ATPV 11.2 CAL/CM2 (HAF = 80.4%)).
Spécifications
- Catalog No:
- 76105-644
- 76105-648
- 76105-646
- 76105-660
- 76105-658
- 76105-570
- 76105-664
- 76105-662
- 76105-652
- 76105-564
- 76105-650
- 76105-656
- 76105-568
- 76105-654
- 76105-566
- 76105-674
- 76105-668
- 76105-666
- 76105-672
- 76105-670
- Model:
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- UBIZ5
- Garment application:
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Fabric:
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish