Bizweld FR Coveralls, Portwest
Fournisseur: Portwest
Certificats
A propos de cet article
Coveralls are designed to protect against radiant, convective, and contact heat.
- Pockets: two front, two back, one concealed cell phone pocket, two side, and one rule
- Dual hazard protection
- ARC2
Certified protection against molten metal splash and guaranteed flame resistance for the life of the garment.
Certifications: EN ISO 11612 (A1+A2. B1. C1. E1. F1). EN ISO 11611 Class 1 (A1+A2). NFPA® 70E. ASTM F1506-10a. NFPA® 2112. ASTM F1959/F1959M-12 (ATPV 11.2 CAL/CM2 (HAF = 80.4%)).
Spécifications
- Catalog No:
- 76105-642
- 76105-560
- 76105-558
- 76105-640
- 76105-638
- 76105-528
- 76105-526
- 76105-532
- 76105-530
- 76105-520
- 76105-562
- 76105-524
- 76105-522
- 76105-556
- 76105-538
- 76105-550
- 76105-636
- 76105-548
- 76105-554
- 76105-552
- 76105-542
- 76105-540
- 76105-546
- 76105-544
- 76105-536
- 76105-534
- Model:
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- UBIZ1
- Garment application:
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Industrial, Fire protection
- Risk area:
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Flame-retardant
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish
- Bizweld - 100% cotton, FR finish