Unit Atoms, Bonds, and Accessories
Fournisseur: Cochranes of Oxford
Certificats
A propos de cet article
Order extra atoms, bonds, and accessories to supplement your unit sets or to build custom models.
- Plastic grey bonds and alloy bonds are fit over prongs to connect spheres
- Flexible white bonds fit over prongs to connect spheres
Large atom spheres have holes which pegs are pushed into to make prongs at all required angles. Different shaped pegs connect spheres and bonds at the correct angles.
Customers Who Bought This Also Bought