LD-100 System Frocks, 470/478 Series, WORKLON®
Fournisseur: Superior Uniform
Certificats
A propos de cet article
Snap closure, set-in sleeve frocks are manufactured from 100% polyester.
- Adjustable snaps at wrists and collar
- Color-coded hang loop denoting size
- Unisex design in a wide range of sizes
Taffeta can be used in a variety of industries requiring particulate control and resistance to chemicals and abrasion. These garments are gamma compatible and autoclavable.
Certifications: Engineered to meet up to Class 1000 (ISO 6) cleanroom requirements.
Spécifications
- Catalog No:
- 10830-542
- 10830-538
- 10830-540
- 10831-029
- 10831-018
- 10831-040
- 10831-030
- 10831-084
- 10831-073
- 10827-212
- 10827-210
- 10831-095
- 10830-508
- 10830-536
- 10830-510
- 10831-007
- Garment application:
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Industrial, Chemical protection
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester