LD-100 System Frocks, 3508/3509/3510 Series, WORKLON®
Fournisseur: Superior Uniform
Certificats
A propos de cet article
Snap closure, raglan sleeve frocks feature polyester knit cuffs and two-ply reinforced placket.
- Adjustable snaps at collar
- Color-coded hang loop denoting size
- Unisex design in a wide range of sizes
Garments are manufactured from 99% polyester, 1% carbon yarn Maxima® high density ESD polyester with a carbon yarn stripe. This fabric can be used in a variety of industries including pharmaceutical, biotechnical, biological, and food processing. Ideal for applications where particulate control, and fluid and bacteria resistance is essential.
Certifications: Engineered to meet up to Class 10 (ISO 4) cleanroom requirements.
Spécifications
- Catalog No:
- 89136-738
- 89136-760
- 89136-740
- 89136-748
- 89136-750
- 89136-744
- 89136-766
- 89136-746
- 89136-768
- 89136-734
- 89136-756
- 89136-736
- 89136-758
- 89136-752
- 89136-732
- 89136-754
- 89136-762
- 89136-742
- 89136-764
- Garment application:
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Medical, Food, Industrial
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Closure type:
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Buttons
- Fabric:
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester
- 100% polyester