Backyard Brains® DIY Muscle Spikerbox
# de catalogue 470323-210
Fournisseur: BACKYARD BRAINS, INC.
Spécifications
- Description:DIY Muscle Spikerbox
- Cat. no.:470323-210
Spécifications
A propos de cet article
Great student project.
- Shipped as individual components for your students to assemble
- Estimated time from 1.5 (hobbyist) to 4 (novice) hours
The muscle spiker box is now available in kit form to record EMGs. The kit takes ~4 hours (complete novice) to 1.5 hours (electronics hobbyist) to build. To build the kit, you will need a soldering iron, some solder, and wire cutters. A small magnifying glass to read the labels on the chips may be helpful for those of you with some mileage.
Customers Who Bought This Also Bought