Stainless Steel Bristle and Wood Shoe Handle Scratch Brush, Gordon Brush®
# de catalogue 22940-644
Fournisseur: Gordon Brush
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Bristle Material:Stainless steel
- Brush Head Diameter:0.006"
- Handle Material:Wood
- Description:Scratch Brush
- Brush Head Width:3/4"
- Cat. no.:22940-644
- No. of Rows:4×16
- Handle Length:10"
- Width:11/8"
- Supplier No.:444SS006G
- Brush Head Length:4 3/4"
- Trim Length:1⅛"
Spécifications
A propos de cet article
This Plater's Brush is made with fine fill material, and recommended for use on plated parts or fine metal finishing.
- Contoured for a comfortable grip
Hardwood shoe handle.
Customers Who Bought This Also Bought