Sable Shader Artist Brush, Gordon Brush
# de catalogue 93001-852
Fournisseur: Gordon Brush
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Bristle Material:Sable hair
- Handle Material:Wood
- Description:Artist Brush
- Size:# 2
- Total length:7"
- Brush Head Width:1/8"
- Cat. no.:93001-852
- Supplier No.:0611-02000
- Trim Length:¼"
Spécifications
A propos de cet article
This Sable Shader Brush is a traditional Flat Brush for applying paints and other liquids to a flat, wide area.
- Metallic turquoise lacquered handle
- Seamless nickel-plated ferrule
- Wood handle
- Flat bristle trim
Made with pure red sable.
Customers Who Bought This Also Bought