Hog Bristle and Plastic Handle Sash Brush, Gordon Brush
# de catalogue 22940-750
Fournisseur: Gordon Brush
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Bristle Material:Hog hair
- Handle Material:Plastic
- Description:Parts Cleaning Brush
- Size:# 6
- Total length:101/8"
- Brush Head Width:11/4"
- Cat. no.:22940-750
- Supplier No.:900162
- Trim Length:2"
Spécifications
A propos de cet article
This Size 6 hog bristle and plastic handle sash brush has an oval shape, and is excellent for dusting and removing debris.
- Features a metal ferrule
Large tapered plastic handle.
Customers Who Bought This Also Bought