Nylon Bristle and Tin Handle Acid Brushes, Gordon Brush®
Fournisseur: Gordon Brush
Certificats
A propos de cet article
These disposable acid brushes are very popular for one-time use.
- Disposable acid brush with Nylon bristle
- Rolled tin handle
Ideal for applying acid flux, glue, lubrication, and other coatings or using for small-area cleaning and the removal of debris.
Customers Who Bought This Also Bought