Tampico Bristle and Wood Shoe Handle Scratch Brush, Gordon Brush
# de catalogue 22940-634
Fournisseur: Gordon Brush
Product available on GSA Advantage®
Small Business Enterprise
Spécifications
- Bristle Material:Tampico
- Handle Material:Wood
- Description:Plater Brush
- Brush Head Width:3/4"
- Cat. no.:22940-634
- No. of Rows:4×16
- Handle Length:10"
- Width:11/8"
- Supplier No.:444T
- Brush Head Length:43/4"
- Trim Length:1"
Spécifications
A propos de cet article
This Plater's Brush is made with fine fill material, and recommended for use on plated parts or fine metal finishing.
- Hardwood shoe handle
Contoured for a comfortable grip.
Customers Who Bought This Also Bought