3419 Series 41" Snap Front Lab Coats, Fashion Seal Healthcare®
Supplier: Superior Uniform
Certificates
About this item
Reusable snap front lab coats are manufactured from an 80% polyester, 20% cotton poplin blend.
- Snap front
- No pockets
- White Polyester knit cuffs
- Convertible collar
- No side slits
Snap front lab coats are ideally suited for use in general-purpose, food, and laboratory applications. Coats are 104.1 cm (41") in length based on size medium.
Specifications
- Catalog No:
- 75817-704
- 75817-718
- 75817-716
- 75817-714
- 75817-712
- 75817-710
- 75817-708
- 75817-706
- Model:
- 3419
- 3419
- 3419
- 3419
- 3419
- 3419
- 3419
- 3419
- Garment application:
- Food, General Purpose, Laboratory
- Food, General Purpose, Laboratory
- Food, General Purpose, Laboratory
- Food, General Purpose, Laboratory
- Food, General Purpose, Laboratory
- Food, General Purpose, Laboratory
- Food, General Purpose, Laboratory
- Food, General Purpose, Laboratory
- Gender:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Closure type:
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Press studs / snaps
- Fabric:
- 80% polyester, 20% cotton poplin blend
- 80% polyester, 20% cotton poplin blend
- 80% polyester, 20% cotton poplin blend
- 80% polyester, 20% cotton poplin blend
- 80% polyester, 20% cotton poplin blend
- 80% polyester, 20% cotton poplin blend
- 80% polyester, 20% cotton poplin blend
- 80% polyester, 20% cotton poplin blend