Biohazardous Waste Bags, Medegen Medical Products
Fournisseur: Medegen Medical Products, LLC
Certificats
A propos de cet article
These biohazardous waste bags are manufactured to meet the specific performance standards in accordance with Federal and specific State Guidelines.
- Biohazardous labeling in both English and Spanish
- Latex-free and environmentally safe for incineration disposal
Premium LLDPE liners ensure consistent strength and superior performance.
Certifications: Bags meet both ASTM D1922 (Elmendorf Tear 480g) and ASTM D1709 Dart Impact (160g) requirements. Complies with US DOT interstate transport of biohazardous waste and all 50 states.
Customers Who Bought This Also Bought