R10000 M611, Printer Ribbons for BMP®61 Labeling System BradyPrinter™, Brady
Fournisseur: Brady Worldwide
Certificats
A propos de cet article
Switch out ribbons in seconds with these easy-to-load ribbon cartridges that slide into place in the always-correct direction.
- Use with BradyPrinter™ M611 and BMP® 61 labeling system
- Single color ribbon
Durable R10000 series ribbons are based on a resin formulation that produces long-lasting printed text that can withstand years in industrial areas and outdoor conditions. These self-contained ribbon cartridges are easy to replace, eliminating the hassle of printer calibration and ribbon threading. These ribbons are ideal for use with long-lasting indoor/outdoor materials such as Brady B-595.
Customers Who Bought This Also Bought