ContactUsLinkComponent
Endure™ Aqueous Clarifier, Cancer Diagnostics
Catalogue # 10809-898
Fournisseur:  Avantor
Numéro CAS:  
undefined
Endure™ Aqueous Clarifier, Cancer Diagnostics
Catalogue # 10809-898
Fournisseur:  Avantor
Numéro CAS:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Spécifications
  • Description:
    Endure™ Aqueous Clarifier
  • Size:
    500 ml
  • Cat. no.:
    10809-898
  • Supplier No.:
    ED44
Spécifications
À propos de cet article
The Endure Staining System is designed to produce precise, consistent H&E staining.
  • Focus clarifier for background haze and stain removal
Staining times in hematoxylin and eosin will depend on the use and the personal preference of the diagnosing physician. CDI has guidelines that will help establish a custom protocol for your specific needs. The Endure H&E System includes Hematoxylin ED1, (or ED5 “XT” for darker staining), Eosin ED2, Bluing ED3 and Clarifier (Aqueous) ED4 or Clarifier (Alcoholic) ED6. Each component is analytically optimized for staining consistency and longevity