A propos de cet article
CF®DI dye hydrazides react with molecules containing aldehyde or ketone groups to form a hydrazone bond. The dyes are size- and charge-matched and do not significantly alter the molecular weight or pI of the labeled proteins. They are used for detection of protein carbonylation by fluorescence 2D-DIGE.
- React with carbonyl moieties of aldehyde or ketone groups.
- Size- and charge-matched, do not significantly alter the molecular weight or pI of the labeled protein.
- For detection of protein carbonylation by fluorescence 2D-DIGE.
- Available in several colors for detection of multiple samples on the same gel.
Also see our regular CF® Dye Hydrazides, available in a wide selection of colors.