Leybold Basic Science Kits, Biology
Supplier: VWR International
Certificates
About this item
Tough, simple, affordable.
- Experiment configurations and instructions adjusted to the curricula
- Basic experiments literature on CD and available from the document center
- Suitable CASSY solution
- Storage case fitting the shape of the apparatus in plastic box with lid
Base equipment set with stand material for basic experiments in botany and ecology.
Principles equipment set with stand material for experiments in microbiology and sense physiology and for experiments for botany and ecology together with the equipment set science kit basis.
Genetics equipment set with for basic experiments in microscopy and genetics.
Customers Who Bought This Also Bought