A propos de cet article
Grinding mill with powerful motor, which is suitable for all laboratory dry milling applications.
- Powerful motor provides speeds up to 6000 min-1 with low noise emissions (less than 70 dB(A) at 6000 min-1 without load) due to brushless 3 phase AC motor
- Milling chamber can be equipped with hammer grinding or blade grinding attachment (second milling chamber not required)
- Integrated speed controller with LED display shows speed and status messages
- Integrated overheating, overload and blocking protection means motor shuts down if overload conditions arise, ensuring the motor is protected
- Safety switch for automatic switch-off, and energy saving automatic standby mode
Milling chamber with attached funnel and rotary slide for sample feed, can be used with interchangeable hammer or blade grinding attachments, and sieves with mesh sizes from 0.2 up to 6 mm. Chamber parts in contact with the sample are made from hardened stainless steel, the anodized aluminum base frame has an integrated handle. The ground material can be collected using vessels with NS 29/32 standard connections (holding clip necessary) or receiving tubes.
This laboratory mill is characterized by a very robust design using solid milled parts and hardened components that have been designed for high wear and tear found with such processes as grinding of stones which can have a hardness of up to 6 on the Mohs scale. You will receive tried and tested Swiss quality.
Spécifications
- Catalog No:
- 76359-402
- 76359-404
- Final fineness:
- < 40 µm (Attainable particle sizes, depending on product)
- < 40 µm (Attainable particle sizes, depending on product)
- Mesh size:
- 0.2…6 mm
- 0.2…6 mm
- Dimensions:
- 325L×251W×480H mm
- 325L×251W×480H mm
- Hopper volume/throughput:
- 300 ml
- 300 ml
- Motor power:
- 1000 W, 115 V
- 1000 W, 115 V
- Sound level:
- 70 dB at 6000 rpm
- 70 dB at 6000 rpm
- Weight:
- 12 kg
- 12 kg