Cleansing Enema Sets, Medegen Medical Products
Fournisseur: Medegen Medical Products, LLC
Certificats
A propos de cet article
Convenient prepackaged sets for administering enemas.
- Bucket and bag type
- Holds 1500 ml of fluid
Other components: 60" tubing with pre-lubricated tip, adjustable clamp, castile soap and waterproof drape.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.