Spouted Collection Containers, Medegen Medical Products
Fournisseur: Medegen Medical Products, LLC
Certificats
A propos de cet article
These 8 oz. spouted containers are available in both polystyrene and polypropylene. Both options come with a convenient pour spout for simplified specimen transfer.
- Graduated in 0.5 oz. and 10 cc (ml) increments
- Polystyrene provides crystal clear clarity
- Polypropylene (translucent) provides improved durability
Ordering information: Printed lids can be purchased separately.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.