Sharps-Tainer ™ Chemotherapy Sharp Containers, Medegen Medical Products
Supplier: Medegen Medical Products, LLC
Certificates
About this item
Heavy duty, non-stackable, sharps-Tainers ™ are ideal for the containment and disposal of chemotherapeutic waste.
- Durable polypropylene construction provides a stable base during use
- Funneled vertical drop and extended neck design help minimize needle exposure and overfilling
- Chemotherapy waste labeling/CYTA for easy identification
Delivery: Supplied with standard locking cap.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.