American Line Two Sided Film Slitter, AccuTec Blades, Inc.
# de catalogue 76339-900
Fournisseur: Accutec Blades
Spécifications
- Description:Film Slitter
- Cat. no.:76339-900
Spécifications
A propos de cet article
These disposable film slitters cut film and open boxes with ease and reduce risk of damage or injury.
- Recessed blade design prevents injuries
- Blunt steel punch on the end punches through box tape, helping with the initial cut
- Two blade surfaces, works great for left- or right-handed users
Carbon steel blade for strength and durability.
Customers Who Bought This Also Bought